Merck
Lubiprostone stimulates small intestinal mucin release.
BMC gastroenterology (2012-11-08)
Robert C De Lisle
ABSTRACT
Lubiprostone is a synthetic bicyclic fatty acid derivative of prostaglandin E1 (PGE1) used for chronic constipation. The best known action of lubiprostone is simulation of Cl- dependent fluid secretion. In a mouse model of the genetic disease cystic fibrosis, we previously showed that in vivo administration of lubiprostone resulted in greater mucus accumulation in the small intestine. The aim of this study was to directly test whether lubiprostone stimulates intestinal mucin release. Mucin release was measured by mounting segments (4-5 cm) of mouse proximal-mid small intestine in an organ bath, allowing access to the perfusate (luminal) and the bath (serosal) solutions. Nifedipine (10-6 M) and indomethacin (10-5 M) were included in all solutions to inhibit smooth muscle activity and endogenous prostaglandin production, respectively. The tissue was equilibrated under flow for 30 min, using the perfusate collected during the final 10 min of the equilibration period to measure unstimulated release rate. Stimulus was then added to either the perfusate or the bath and the perfusate was collected for another 30 min to measure the stimulated mucin release rate. Mucin in perfusates was quantified by periodic acid-Schiff's base dot-blot assay, using purified pig gastric mucin as a standard. When applied luminally at 1 μM lubiprostone was ineffective at stimulating mucin release. When added to the serosal solution, 1 μM lubiprostone stimulated mucin release to ~300% of the unstimulated rate. As a positive control, serosal 1 μM prostaglandin E2 increased mucin release to ~400% of the unstimulated rate. These results support the idea that lubiprostone has prostaglandin-like actions on the intestine, which includes stimulation of mucin release. Stimulation of mucin release by lubiprostone may be protective in gastrointestinal conditions where loss of mucus is believed to contribute to pathogenesis. Thus, in addition to chronic constipation, there is greater potential for the therapeutic applications of lubiprostone.
Sigma-Aldrich
Periodic Acid-Schiff (PAS) Staining System
Sigma-Aldrich
Prostaglandin E1, ≥98% (HPLC), synthetic
Sigma-Aldrich
Prostaglandin E1, synthetic, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Prostaglandin E1, powder, γ-irradiated, BioXtra, suitable for cell culture
Alprostadil, European Pharmacopoeia (EP) Reference Standard