Merck
Pattern hair loss in men: diagnosis and medical treatment.
Dermatologic clinics (2012-11-20)
Nusrat Banka, M J Kristine Bunagan, Jerry Shapiro
ABSTRACT
Androgenetic alopecia is a common cause of hair loss in both men and women. The exact pathogenesis of androgenetic alopecia is not well understood. As the name implies, the role of androgens and genetic susceptibility predisposes to pattern hair loss due to gradual conversion of terminal hair into vellus hair. Male and female pattern hair loss are clinically distinct entities but histologically indistinguishable. The role of sex hormones in females is less understood. This article discusses current understanding of the etiopathogenesis of hair loss in men, diagnostic tests available, and its medical management.
Sigma-Aldrich
Minoxidil, ≥99% (TLC)
Minoxidil, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Finasteride, ≥98% (HPLC), powder