Merck
Solubility-pH profiles of some acidic, basic and amphoteric drugs.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences (2012-11-28)
Elham Shoghi, Elisabet Fuguet, Elisabeth Bosch, Clara Ràfols
ABSTRACT
The solubility vs. pH profiles of five ionizable drugs of different nature (a monoprotic acid, a monoprotic base, a diprotic base and two amphoteric compounds showing a zwitterionic species each one) have been determined through two different methodologies: the classical shake-flask (S-F) and the potentiometric Cheqsol methods using in both instances the appropriate Henderson-Hasselbalch (H-H) or derived relationships. The results obtained independently from both approaches are consistent. A critical revision about the influence of the electrolyte used as buffering agent in the S-F method on the obtained solubility values is also performed. Thus, some deviations of the experimental points with respect the H-H profiles can be attributed to specific interactions between the buffering electrolyte and the drug due to the hydrotrophic character of citric and lactic acids. In other cases, the observed deviations are independent of the buffers used since they are caused by the formation of new species such as drug aggregates (cefadroxil) or the precipitation of a salt from a cationic species of the analyzed compound (quetiapine).
Sigma-Aldrich
Propyl 4-hydroxybenzoate, ≥99%
Sigma-Aldrich
Propyl 4-hydroxybenzoate, tested according to Ph. Eur.
Supelco
Cefadroxil, analytical standard
Sigma-Aldrich
Propyl 4-hydroxybenzoate, BioXtra
Supelco
Acebutolol hydrochloride, analytical standard
Supelco
Sulfadimethoxine, 98.0-102.0%
Supelco
Sulfadimethoxine, VETRANAL®, analytical standard
Supelco
Propylparaben, Pharmaceutical Secondary Standard; Certified Reference Material
Propyl parahydroxybenzoate, European Pharmacopoeia (EP) Reference Standard
Acebutolol hydrochloride, European Pharmacopoeia (EP) Reference Standard