Merck
  • Efficient copper-catalyzed Michael addition of acrylic derivatives with primary alcohols in the presence of base.
Efficient copper-catalyzed Michael addition of acrylic derivatives with primary alcohols in the presence of base.
Chemical communications (Cambridge, England) (2012-11-30)
Feng Wang, Haijun Yang, Hua Fu, Zhichao Pei
ABSTRACT
A novel and efficient copper-catalyzed Michael addition of acrylic derivatives with primary alcohols in the presence of base has been developed. The protocol uses readily available acrylic derivatives and primary alcohols as the starting materials, inexpensive CuCl(2) as the catalyst, and the corresponding addition products were obtained in moderate to excellent yields.
Sigma-Aldrich
Copper(II) chloride dihydrate, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Copper(II) chloride dihydrate, ≥99.95% trace metals basis
Sigma-Aldrich
Copper(II) chloride dihydrate, 99.999%
Sigma-Aldrich
Copper(II) chloride dihydrate, ACS reagent, ≥99.0%
Sigma-Aldrich
Copper(II) chloride dihydrate, reagent grade
Sigma-Aldrich
Copper(II) chloride, 99.999% trace metals basis
Sigma-Aldrich
Copper(II) chloride, anhydrous, powder, ≥99.995% trace metals basis
Sigma-Aldrich
Copper(II) chloride, 97%
Sigma-Aldrich
Copper(II) chloride, powder, 99%