Merck
PEM anchorage on titanium using catechol grafting.
PloS one (2012-12-12)
Hélène Marie, Amélie Barrere, Frederic Schoenstein, Frédérique Schoentstein, Marie-Hélène Chavanne, Brigitte Grosgogeat, Laurence Mora
ABSTRACT
This study deals with the anchorage of polyelectrolyte films onto titanium surfaces via a cathecol-based linker for biomedical applications. The following study uses a molecule functionalized with a catechol and a carboxylic acid: 3-(3,4-dihydroxyphenyl)propanoic acid. This molecule is anchored to the TiO(2) substrate via the catechol while the carboxylic acid reacts with polymers bearing amine groups. By providing a film anchorage of chemisorption type, it makes possible to deposit polyelectrolytes on the surface of titanium. Infrared spectroscopy (ATR-FTIR), X-ray photoelectron spectroscopy (XPS), contact angle and atomic force microscopy (AFM) measurements show that the different steps of grafting have been successfully performed. This method based on catechol anchorage of polyelectrolytes open a window towards large possibilities of clinical applications.
Sigma-Aldrich
Pyrocatechol, ≥99%
Sigma-Aldrich
Pyrocatechol, purified by sublimation, ≥99.5%
Sigma-Aldrich
3,4-Dihydroxyhydrocinnamic acid, 98%
Sigma-Aldrich
1,2-Dihydroxybenzene, ReagentPlus®, ≥99%