Merck
  • Microfluidic free-flow electrophoresis chips with an integrated fluorescent sensor layer for real time pH imaging in isoelectric focusing.
Microfluidic free-flow electrophoresis chips with an integrated fluorescent sensor layer for real time pH imaging in isoelectric focusing.
Chemical communications (Cambridge, England) (2012-12-19)
Stefan Jezierski, Detlev Belder, Stefan Nagl
ABSTRACT
Functional microfluidic free-flow electrophoresis chips with integrated fluorescent pH sensors are presented. Polyethylene glycol based structures were fabricated that allowed for integration of both functions on a single microchip. Microchips were applied in free-flow isoelectric focusing of model compounds and proteins with on-line monitoring of pH during microscale electrophoretic separation.
Sigma-Aldrich
Conalbumin from chicken egg white, Iron complex
Sigma-Aldrich
Conalbumin from chicken egg white, Substantially iron-free
Sigma-Aldrich
Conalbumin from chicken egg white, BioReagent, suitable for cell culture