Merck
  • Stability and antioxidant activity of semi-synthetic derivatives of 4-nerolidylcatechol.
Stability and antioxidant activity of semi-synthetic derivatives of 4-nerolidylcatechol.
Molecules (Basel, Switzerland) (2012-12-25)
Emerson Silva Lima, Ana Cristina Silva Pinto, Karla Lagos Nogueira, Luiz Francisco Rocha e Silva, Patricia Danielle Oliveira de Almeida, Marne Carvalho de Vasconcellos, Francisco Celio Maia Chaves, Wanderli Pedro Tadei, Adrian Martin Pohlit
ABSTRACT
4-nerolidylcatechol (4-NC) is an unstable natural product that exhibits important antioxidant, anti-inflammatory and other properties. It is readily obtainable on a multi-gram scale through straightforward solvent extraction of the roots of cultivated Piper peltatum or P. umbellatum, followed by column chromatography on the resulting extract. Semi-synthetic derivatives of 4-NC with one or two substituent groups (methyl, acetyl, benzyl, benzoyl) on the O atoms have been introduced that have increased stability compared to 4-NC and significant in vitro inhibitory activity against the human malaria parasite Plasmodium falciparum. Antioxidant and anti-inflammatory properties may be important for the antiplasmodial mode of action of 4-NC derivatives. Thus, we decided to investigate the antioxidant properties, cytotoxicity and stability of 4-NC derivatives as a means to explore the potential utility of these compounds. 4-NC showed high antioxidant activity in the DPPH and ABTS assays and in 3T3-L1 cells (mouse embryonic fibroblast), however 4-NC was more cytotoxic (IC₅₀ = 31.4 µM) and more unstable than its derivatives and lost more than 80% of its antioxidant activity upon storage in solution at -20 °C for 30 days. DMSO solutions of mono-O-substituted derivatives of 4-NC exhibited antioxidant activity and radical scavenging activity in the DPPH and ABTS assays that was comparable to that of BHA and BHT. In the cell-based antioxidant model, most DMSO solutions of derivatives of 4-NC were less active on day 1 than 4-NC, quercetin and BHA and more active antioxidants than BHT. After storage for 30 days at -20 °C, DMSO solutions of most of the derivatives of 4-NC were more stable and exhibited more antioxidant activity than 4-NC, quercetin and BHA and exhibited comparable antioxidant activity to BHT. These findings point to the potential of derivatives of 4-NC as antioxidant compounds.
Sigma-Aldrich
2,6-Di-tert-butyl-4-methylphenol, tested according to Ph. Eur.
Supelco
3,5-Di-tert-butyl-4-hydroxytoluene, analytical standard
Butylhydroxytoluene, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Butylated hydroxyanisole, 99%, FCC, FG
Sigma-Aldrich
2,6-Di-tert-butyl-4-methylphenol, purum, ≥99.0% (GC)
Sigma-Aldrich
2,6-Di-tert-butyl-4-methylphenol, ≥99.0% (GC), powder
Sigma-Aldrich
Butylated hydroxyanisole, ≥98.5%
Supelco
Butylated Hydroxytoluene, Pharmaceutical Secondary Standard; Certified Reference Material
Butylhydroxyanisole, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Butylated hydroxytoluene, ≥99%, FCC, FG