Merck
  • Complete dechlorination of 2,4-dichlorophenol in aqueous solution on palladium/polymeric pyrrole-cetyl trimethyl ammonium bromide/foam-nickel composite electrode.
Complete dechlorination of 2,4-dichlorophenol in aqueous solution on palladium/polymeric pyrrole-cetyl trimethyl ammonium bromide/foam-nickel composite electrode.
Journal of hazardous materials (2012-12-29)
Zhirong Sun, Xuefeng Wei, Yanbo Han, Shan Tong, Xiang Hu
ABSTRACT
The electrochemically reductive dechlorination of 2,4-dichlorophenol (2,4-DCP) in aqueous solution on palladium/polymeric pyrrole-cetyl trimethyl ammonium bromide/foam-nickel electrode (Pd/PPy-CTAB/foam-Ni electrode) was investigated in this paper. Pd/PPy-CTAB/foam-Ni electrode was prepared and characterized by cyclic voltammetry (CV), scanning electron microscope (SEM), X-ray diffraction (XRD), Brunauer-Emmett-Teller (BET) adsorption and inductively coupled plasma-atomic emission spectroscopy (ICP-AES). The influences of some experimental factors such as the dechlorination current, dechlorination time and the initial pH on the removal efficiency and the current efficiency of 2,4-DCP dechlorination on Pd/PPy-CTAB/foam-Ni electrode were studied. Complete removal of 2,4-DCP was achieved and the current efficiency of 47.4% could be obtained under the conditions of the initial pH of 2.2, the dechlorination current of 5 mA and the dechlorination time of 50 min when the initial 2,4-DCP concentration was 100 mg L(-1). The analysis of high performance liquid chromatography (HPLC) identified that the intermediate products were 2-chlorophenol (2-CP) and 4-chlorophenol (4-CP). The final products were mainly phenol. Its further reduction product cyclohexanone was also detected. The electrocatalytic dechlorination pathways of 2,4-DCP on Pd/PPy-CTAB/foam-Ni electrode were discussed. The stability of the electrode was favorable that it could keep dechlorination efficiency at 100% after having been reused 10 times. Results revealed that the stable prepared Pd/PPy-CTAB/foam-Ni electrode presented a good application prospect in dechlorination process with high effectiveness and low cost.
Product Description
Supelco
Hexadecyltrimethylammonium bromide, analytical standard
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥98%
Supelco
Hexadecyltrimethylammonium bromide, suitable for ion pair chromatography, LiChropur
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥96.0% (AT)
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioXtra, ≥99%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioUltra, Molecular Biology, ≥99.0% (AT)
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, Molecular Biology, ≥99%
Supelco
Hexadecyltrimethylammonium bisulfate, suitable for ion pair chromatography, LiChropur, ≥99.0% (T)
Sigma-Aldrich
Hexadecyltrimethylammonium hydroxide solution, 10 wt. % in H2O
Sigma-Aldrich
Hexadecyltrimethylammonium chloride, ≥98.0% (NT)
Sigma-Aldrich
Cetyltrimethylammonium chloride solution, 25 wt. % in H2O
Sigma-Aldrich
Hexadecyltrimethylammonium hydroxide solution, ~25% in methanol (T)
Sigma-Aldrich
Cetyltrimethylammonium hydrogensulfate, 99%
Sigma-Aldrich
2,4-Dichlorophenol, 99%