Merck
  • Preparation, preliminary screening of new types of steroid conjugates and their activities on steroid receptors.
Preparation, preliminary screening of new types of steroid conjugates and their activities on steroid receptors.
Steroids (2013-01-08)
Michal Jurášek, Petr Džubák, David Sedlák, Hana Dvořáková, Marián Hajdúch, Petr Bartůněk, Pavel Drašar
ABSTRACT
There were synthesized new types of ribbon type steroidal dimers derived from three types of steroidal skeletons (cholic acid, etienic acid, estrone) using Cu(I) catalyzed 1, 3-dipolar cycloaddition reaction. Steroid parts of the molecular "ribbons" are linked by heterocyclic moiety, namely by 2,6-bis((1H-1,2,3-triazol-1-yl)-methyl)pyridine. Compounds synthesized possess different cytotoxic and hormone receptor modulating activities.
Product Description
Supelco
Estrone solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Estrone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Estrone, ≥99%
Sigma-Aldrich
Cholic acid, from bovine and/or ovine, ≥98%
Cholic acid, European Pharmacopoeia (EP) Reference Standard