Merck
  • Emodin regulates glucose utilization by activating AMP-activated protein kinase.
Emodin regulates glucose utilization by activating AMP-activated protein kinase.
The Journal of biological chemistry (2013-01-11)
Parkyong Song, Jong Hyun Kim, Jaewang Ghim, Jong Hyuk Yoon, Areum Lee, Yonghoon Kwon, Hyunjung Hyun, Hyo-Youl Moon, Hueng-Sik Choi, Per-Olof Berggren, Pann-Ghill Suh, Sung Ho Ryu
ABSTRACT
AMP-activated protein kinase has been described as a key signaling protein that can regulate energy homeostasis. Here, we aimed to characterize novel AMP-activated kinase (AMPK)-activating compounds that have a much lower effective concentration than metformin. As a result, emodin, a natural anthraquinone derivative, was shown to stimulate AMPK activity in skeletal muscle and liver cells. Emodin enhanced GLUT4 translocation and [(14)C]glucose uptake into the myotube in an AMPK-dependent manner. Also, emodin inhibited glucose production by suppressing the expression of key gluconeogenic genes, such as phosphoenolpyruvate carboxykinase and glucose-6-phosphatase, in hepatocytes. Furthermore, we found that emodin can activate AMPK by inhibiting mitochondrial respiratory complex I activity, leading to increased reactive oxygen species and Ca(2+)/calmodulin-dependent protein kinase kinase activity. Finally, we confirmed that a single dose administration of emodin significantly decreased the fasting plasma glucose levels and improved glucose tolerance in C57Bl/6J mice. Increased insulin sensitivity was also confirmed after daily injection of emodin for 8 days using an insulin tolerance test and insulin-stimulated PI3K phosphorylation in wild type and high fat diet-induced diabetic mouse models. Our study suggests that emodin regulates glucose homeostasis in vivo by AMPK activation and that this may represent a novel therapeutic principle in the treatment of type 2 diabetic models.
Sigma-Aldrich
Emodin, from Frangula bark, ≥90% (HPLC)
Supelco
Emodin, analytical standard