Merck
  • The protective effect of glycyrrhetinic acid on carbon tetrachloride-induced chronic liver fibrosis in mice via upregulation of Nrf2.
The protective effect of glycyrrhetinic acid on carbon tetrachloride-induced chronic liver fibrosis in mice via upregulation of Nrf2.
PloS one (2013-01-24)
Shaoru Chen, Liyi Zou, Li Li, Tie Wu
ABSTRACT
This study was designed to investigate the potentially protective effects of glycyrrhetinic acid (GA) and the role of transcription factor nuclear factor-erythroid 2(NF-E2)-related factor 2 (Nrf2) signaling in the regulation of Carbon Tetrachloride (CCl(4))-induced chronic liver fibrosis in mice. The potentially protective effects of GA on CCl(4)-induced chronic liver fibrosis in mice were depicted histologically and biochemically. Firstly, histopathological changes including regenerative nodules, inflammatory cell infiltration and fibrosis were induced by CCl(4).Then, CCl(4) administration caused a marked increase in the levels of serum aminotransferases (GOT, GPT), serum monoamine oxidase (MAO) and lipid peroxidation (MDA) as well as MAO in the mice liver homogenates. Also, decreased nuclear Nrf2 expression, mRNA levels of its target genes such as superoxide dismutase 3 (SOD3), catalase (CAT), glutathione peroxidase 2 (GPX2), and activity of cellular antioxidant enzymes were found after CCl(4) exposure. All of these phenotypes were markedly reversed by the treatment of the mice with GA. In addition, GA exhibited the antioxidant effects in vitro by on FeCl(2)-ascorbate induced lipid peroxidation in mouse liver homogenates, and on DPPH scavenging activity. Taken together, these results suggested that GA can protect the liver from oxidative stress in mice, presumably through activating the nuclear translocation of Nrf2, enhancing the expression of its target genes and increasing the activity of the antioxidant enzymes. Therefore, GA may be an effective hepatoprotective agent and viable candidate for treating liver fibrosis and other oxidative stress-related diseases.
Sigma-Aldrich
Iron(II) chloride, 98%
Sigma-Aldrich
Iron(II) chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Iron(II) chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Iron(II) chloride tetrahydrate, 99.99% trace metals basis
Sigma-Aldrich
Iron(II) chloride, AnhydroBeads, −10 mesh, 99.998% trace metals basis
Sigma-Aldrich
18β-Glycyrrhetinic acid, 97%
Sigma-Aldrich
Iron(II) chloride tetrahydrate, ReagentPlus®, 98%
Sigma-Aldrich
Iron(II) chloride tetrahydrate, puriss. p.a., ≥99.0% (RT)