Merck
  • Dynamic lipid lateral segregation driven by lauryl cyclodextrin interactions at the membrane surface.
Dynamic lipid lateral segregation driven by lauryl cyclodextrin interactions at the membrane surface.
Langmuir : the ACS journal of surfaces and colloids (2013-02-16)
Michel Roux, Edward Sternin, Véronique Bonnet, Christophe Fajolles, Florence Djedaíni-Pilard
ABSTRACT
Amphiphilic cyclodextrins, with a cholesterol anchor (βChol) or an aspartic acid moiety esterified by two lauryl acyl chains (βDLC), were designed to combine the inclusion ability of the cyclodextrin cavity with the carrier properties of model membranes. Their insertion in phosphatidylcholine bilayers induces a marked lateral phase separation into a pure lipid phase and a cyclodextrin-rich phase (LCD), organized as a 2D cyclodextrin network stabilized by intermolecular hydrogen bonds between the saccharide headgroups at the membrane surface (Roux, M.; Perly, B.; Djedaïni-Pilard, F. Self-Assemblies of Amphiphilic Cyclodextrins. Eur. Biophys. J.2007, 36, 861-867). We have replaced the dilauryl anchor by a single lauryl chain grafted onto a leucine residue, giving monolauryl-β-cyclodextrin (βMLC), which readily inserts into bilayers of chain-deuterated DMPC-d27. The removal of one lauryl acyl chain leads to a dynamic membrane insertion of this new cyclodextrin derivative, with significant lipid exchange on the deuterium NMR time scale between a loosely packed cyclodextrin-enriched phase (L'CD) and free lipid regions, yielding broadened two-component NMR spectra. Like the LCD phases, the cyclodextrin-enriched L'CD regions remain (partially) fluid below the DMPC-d27 main fluid-to-gel transition but do not undergo a clear transition toward a gel state, as observed at 14.5 °C in the LCD phase induced by the dilauryl derivative. Partially fluid lipids of the βMLC-induced L'CD phase coexist with pure lipids in the Pβ' gel phase with possible exchange between them until all of the lipids undergo a transition toward an Lβ' gel state at around 7 °C. Trimethylated monolauryl-β-cyclodextrins induce only an ordering of the lipid acyl chains just above the main transition, without any lateral phase separation. Similar chain ordering is also observed within the βMLC-induced L'CD phase as a consequence of the deep membrane insertion of the monolauryl nonmethylated cyclodextrin derivative.
Sigma-Aldrich
Sodium dodecanoate, 99-100%
Sigma-Aldrich
Lauric acid, natural, ≥98%, FCC, FG
Sigma-Aldrich
Lauric acid, ≥98%, FCC, FG
Avanti
14:0-14:0-d27 PC, Avanti Research - A Croda Brand 860391P, powder
Avanti
14:0 PC-d54, 1,2-dimyristoyl-d54-sn-glycero-3-phosphocholine, chloroform
Avanti
14:0 PC-d54, 1,2-dimyristoyl-d54-sn-glycero-3-phosphocholine, powder
Supelco
Dodecanoic acid, analytical standard
Sigma-Aldrich
Dodecanoic acid, 98%
Sigma-Aldrich
Dodecanoic acid, ≥99% (GC/titration)