Merck
  • The physical characteristics and emulsification properties of partially dephosphorylated bovine β-casein.
The physical characteristics and emulsification properties of partially dephosphorylated bovine β-casein.
Food chemistry (2013-02-16)
Noel A McCarthy, Alan L Kelly, James A O'Mahony, Mark A Fenelon
ABSTRACT
Bovine β-casein was purified from phosphocasein by rennet coagulation and cold solubilisation from the resultant curd. β-Casein was then dephosphorylated using potato acid phosphatase. Urea-polyacrylamide gel electrophoresis (PAGE) of partially dephosphorylated β-casein showed a number of bands, depending on the final level of phosphorylation. Dephosphorylating β-casein increased its pH of minimum solubility from ∼pH 5 to 5.5 and reduced its net negative charge from -30.8 to -27.0 mV. During the acidification of β-casein solutions, partially dephosphorylated β-casein failed to form a gel, unlike the phosphorylated (i.e., control) β-casein. Use of partially dephosphorylated β-casein to stabilise oil-in-water emulsions resulted in larger fat globules compared to control β-casein, but such globules were less susceptible to aggregation in the presence of 15 or 30 mM CaCl(2). Overall, the dephosphorylation of β-casein resulted in a protein similar to human β-casein in terms of physicochemical functionality, with increased stability against calcium-induced aggregation.
Sigma-Aldrich
Casein, Dephosphorylated from bovine milk, lyophilized powder
Sigma-Aldrich
Phosphatase, Acid from wheat germ, ≥0.4 unit/mg solid
Sigma-Aldrich
Phosphatase, Acid from sweet potato, ammonium sulfate suspension, ≥10.0 units/mg protein (modified Warburg-Christian)
Sigma-Aldrich
Phosphatase, Acid from potato, lyophilized powder, ≥0.5 unit/mg solid
Sigma-Aldrich
Phosphatase, Acid from potato, lyophilized powder, ≥3.0 units/mg solid