Merck
  • In vitro seed germination of economically important edible bamboo Dendrocalamus membranaceus Munro.
In vitro seed germination of economically important edible bamboo Dendrocalamus membranaceus Munro.
Indian journal of experimental biology (2013-02-28)
Jasmine Brar, Manju Anand, Anil Sood
ABSTRACT
An in vitro propagation protocol using mature seeds of D. membranaceus was successfully established. Scarcity of seeds in bamboos because of their long flowering periods and irregular seed set resulting in low viability and germination potential, motivated us to undertake the present study. The effects of sterilants, light conditions, exogenous application of plant growth regulators and temperature in overcoming germination barriers in ageing seeds of bamboo were studied. It was found that HgCl2 (0.1%) along with bleach (15%) was more effective in raising aseptic cultures. Dark conditions, high temperatures around 30 degrees C and soaking of seeds in GA3 solution (50 ppm) overnight stimulated high percent of seed germination with corresponding increase in shoot length (2.7 +/- 0.7 mm) and number of sprouts (2.1 +/- 0.7) per explants during culture initiation. 6-benzylaminopurine acted synergistically with kinetin to give optimum germination rate of 70 +/- 13.9% as compared to 63.13% when used individually. For prolonged maintenance of cultures, 2% sucrose was found to be suitable for promoting photomixotrophic micropropagation. Following this procedure, about 65% survival of plantlets could be achieved during hardening. Biochemically seeds consume starchy endosperm for emergence of radicle which is taken as a sign of germination as also evident from the present study. Loss of viability and vigour after a year was confirmed by Tetrazolium chloride test. Micropropagation protocol developed here will ensure regeneration of large number of plants in a relatively short time. Conclusively, in vitro propagation protocol developed in D. membranaceus using mature seeds as an explants is reported for the first time.
Sigma-Aldrich
Kinetin, BioReagent, suitable for plant cell culture, amorphous powder
Sigma-Aldrich
Kinetin, suitable for plant cell culture, crystalline
Sigma-Aldrich
Kinetin, ≥99.0% (HPLC)
Sigma-Aldrich
Kinetin solution, 1 mg/mL, BioReagent, suitable for plant cell culture
Sigma-Aldrich
Gibberellic acid potassium salt, suitable for plant cell culture, BioReagent, ~95%, ≥50% total GA3 basis
Sigma-Aldrich
Gibberellic acid, BioReagent, suitable for plant cell culture, ≥90% gibberellin A3 basis (of total gibberellins.)
Supelco
Gibberellic acid, PESTANAL®, analytical standard
Sigma-Aldrich
Gibberellic acid, 90% gibberellin A3 basis (HPLC)