Merck
  • Role of extra-cellular fatty acids in vancomycin induced biofilm formation by vancomycin resistant Staphylococcus aureus.
Role of extra-cellular fatty acids in vancomycin induced biofilm formation by vancomycin resistant Staphylococcus aureus.
Pakistan journal of pharmaceutical sciences (2013-03-05)
Zulfiqar Ali Mirani, Nusrat Jamil
ABSTRACT
In the present study a vancomycin resistant Staphylococcus aureus (S. aureus) (VRSA) (Labeled as CP2) was isolated from the blood of a post-operative cardiac patient is described. It harbors a plasmid which carry vanA gene and exhibited low-level vancomycin resistance (MIC 16μg/mL), was sensitive to teicoplanin. It has been observed that sub-lethal dose of vancomycin induced biofilm formation by CP2 on nylon and silicon indwelling. The results divulge new insights into associations between vancomycin induced biofilms and extra-cellular fatty acids. Gas chromatography coupled with mass spectrometry (GC-MS) revealed that biofilm matrix of CP2 contains a variety of saturated and un-saturated fatty acids, especially, diverse species of octadecanoic (C18:0) and octadecenoic acids (C18:1). A large difference in fatty acids composition was noticed in biofilms, isolated from hydrophobic and hydrophilic surfaces. CP2 produced thicker layer of biofilms on hydrophobic silicon and nylon surfaces which contains variety of saturated, un-saturated and cyclic fatty acids. Contrary to this on hydrophilic glass surfaces it produced thinner layer of biofilm which contains only straight chain saturated fatty acids. These fatty acid components seem to play a crucial role in cell-cell communication and in the establishment of biofilms, consequently, advantageous for pathogens to survive in hospital environment under enormous antibiotics pressure.
Sigma-Aldrich
Teicoplanin
Sigma-Aldrich
Stearic acid, ≥95%, FCC, FG
Sigma-Aldrich
Stearic acid, Grade I, ≥98.5% (capillary GC)
Sigma-Aldrich
Stearic acid, reagent grade, 95%
Supelco
Stearic acid, analytical standard
Stearic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Stearic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Stearic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland