Merck
  • Impending corneal perforation after collagen cross-linking for herpetic keratitis.
Impending corneal perforation after collagen cross-linking for herpetic keratitis.
Journal of cataract and refractive surgery (2013-03-26)
Giulio Ferrari, Lorenzo Iuliano, Maurizia Viganò, Paolo Rama
ABSTRACT
Collagen crosslinking (CXL) has been proposed as a treatment for infectious keratitis. Given the insurgence of antibiotic-resistant microorganisms and frequent toxicity of topical medications, CXL may be a potential treatment for corneal infections. However, corneal infection is itself a possible complication of this treatment. We describe a case of severe corneal thinning and melting in a woman who had a CXL procedure as a treatment for herpetic keratitis.
Product Description
Supelco
Riboflavin (B2), analytical standard
Supelco
Riboflavin, Pharmaceutical Secondary Standard; Certified Reference Material
Aciclovir, European Pharmacopoeia (EP) Reference Standard
Supelco
Acyclovir, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
(−)-Riboflavin, from Eremothecium ashbyii, ≥98%
Sigma-Aldrich
(−)-Riboflavin, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98%
Sigma-Aldrich
(−)-Riboflavin, meets USP testing specifications
Sigma-Aldrich
Acycloguanosine, ≥99% (HPLC), powder
Riboflavin, European Pharmacopoeia (EP) Reference Standard