Merck
  • [Nitric oxide modification of hemoglobin oxygen affinity in different conditions of oxygenation].
[Nitric oxide modification of hemoglobin oxygen affinity in different conditions of oxygenation].
Rossiiskii fiziologicheskii zhurnal imeni I.M. Sechenova (2013-05-11)
T L Stepuro, V V Zinchuk
ABSTRACT
The aim of the present study was to investigate the role of nitric oxide (NO) in the regulation of hemoglobin oxygen affinity (HOA) in the presence of different oxygen partial pressure. In this research the effect of NO donors on gas-transport, acid-base balance, HOA indexes, metHb, iron-nitrosylhemoglobin amounts, and total nitrite/nitrate concentration was estimated in vitro. Experimentally, positive correlation was found between NO-dependent shift of HOA and hemoglobin oxygen saturation level. In conclusion, NO is a component of autonomous intraerythrocytic system of HOA regulation, which action is determined by oxygen in the blood. We assume that the physiological significance of such NO action is to maintain aerobic metabolism through optimal blood oxygenation in the pulmonary circulation, on the one hand, and, on the other hand, its compensation under low oxygen tension in the working tissues.
Product Description
Supelco
L-Cysteine hydrochloride monohydrate, Pharmaceutical Secondary Standard; Certified Reference Material
Cysteine hydrochloride monohydrate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
L-Cysteine hydrochloride monohydrate, from non-animal source, suitable for cell culture, meets EP, USP testing specifications
Sigma-Aldrich
L-Cysteine hydrochloride monohydrate, reagent grade, ≥98% (TLC)
Sigma-Aldrich
Hemoglobin human, lyophilized powder
Sigma-Aldrich
Hemoglobin porcine, lyophilized powder
Sigma-Aldrich
Hemoglobin from bovine blood, dried erythrocytes
Sigma-Aldrich
Hemoglobin from bovine blood, lyophilized powder
Sigma-Aldrich
Hemoglobin from bovine blood, suitable for protease substrate, substrate powder
Sigma-Aldrich
L-Cysteine hydrochloride monohydrate, BioUltra, ≥99.0% (RT)
Millipore
Hemoglobin from bovine blood, from bovine blood, powder, for Neisseria spp., pack of 100 g or 500 g, suitable for microbiology
Sigma-Aldrich
L-Cysteine hydrochloride monohydrate, Wacker Chemie AG, 98.5-101.0%
Sigma-Aldrich
L-Cysteine, ≥97%, FG
Sigma-Aldrich
L-Cysteine, 97%
SAFC
L-Cysteine
Sigma-Aldrich
L-Cysteine, BioUltra, ≥98.5% (RT)
Sigma-Aldrich
L-Cysteine, from non-animal source, BioReagent, suitable for cell culture, ≥98%
Supelco
L-Cysteine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Cysteine, Wacker Chemie AG, ≥98.0%