Merck
[Chemical constituents from leaves of Nelumbo nucifera].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2013-06-04)
Xiao-Liang Zhao, Zhi-Min Wang, Xiao-Jun Ma, Wen-Guang Jing, An Liu
ABSTRACT
To study the chemical constituents, twenty-seven compounds were isolated from the 70% ethanol extract from leaves of Nelumbo nucifera by modern chromatographic techniques. Their structures were identified as 10-octacosanol (1), beta-sitosterol (2), 1-undecanol (3), 1-eicosanol (4), daucosterol (5), 6'-hydroxy-4,4'-dimethoxychalcone (6), 3,7,8-trimethoxy-1-hydroxy-xanthone (7), rhamnetin-3-O-beta-D-glucopyranoside (8), chrysoeriol-7-O-beta-D-glucoside (9), quercetin-3-O-beta-D-glucopyranoside (10), quercetin-3-O-alpha-L-rhamnopyranosyl (11), hyperoside (12), quercetin-3-O-rutinoside (13), astragalin (14), isorhamnetin-3-O-alpha-L-rhamnopyranosyl-(1--> 6)-[alpha-D-lyxopyranosyl-(1 --> 2) -beta-D-glucopyranoside] (15), isorhamnetin-3-O-alpha-D-lyxopyranosyl-(1 --> 2) -beta-D-glucopyranoside (16), isorhamnetin-3-O-beta-D-glucopyranoside (17), isorhamnetin-3-O-alpha-L-rhamnopyranosyl-(1 --> 6)-beta-D-glucopyranoside (18), quercetin (19), kaempferol (20), dehydronuciferine (21), roemerine (22), stigmast-7-en-3-O-beta-D-glucopyranoside (23), stigmast-7-en-3beta-ol (24), and benzene-1,2-diol (25) on the basis of spectral data analysis. Compounds 1, 6, 7, 8, 24 and 25 were isolated from this plant for the first time, and compounds 15-18 were isolated from the leaves for the first time. Compounds 6, 8, 10, 11, 13 and 15 showed inhibitory activities against beta amyloid (1-42) by A-beta aggregation method with inhibition rates of (63.99 +/- 24.29)%, (79.61 +/- 4.49)%, (49.96 +/- 12.61)%, (101.19 +/- 8.19)%, (88.41+/-6.76)% and (72.48 +/- 8.97)%, respectively.
Product Description
Supelco
Ethanol, standard for GC
Sigma-Aldrich
Ethanol, purum, secunda spirit, denaturated with 2% 2-butanone, S15, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Sigma-Aldrich
Ethanol
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 2% 2-butanone, F25 MEK1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 4.8% isopropanol, A15 IPA1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 2% 2-butanone, A15 MEK1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethanol, absolute, sales not in Germany, ≥99.8% (vol.)
Supelco
Ethanol-40, 40 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-150, 150 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Supelco
Ethanol-10, 10 mg/dL in H2O, pack of 10 × 1.2 mL ampules, certified reference material, Cerilliant®
Supelco
Ethanol-100 (10 ampules/kit), 100 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-200 (10 ampules/kit), 200 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-80, 80 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-300, 300 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-50, 50 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.005% water
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.003% water
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Supelco
Ethanol-500, 500 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-400 (10 ampules/kit), 400 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-20 (10 ampules/kit), 20 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-25, 25 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol Calibration Kit, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®