Merck
  • Surface plasmon-enhanced light-emission mechanism of Ag-coated ZnO/Al2O3 core/shell nanorod structures.
Surface plasmon-enhanced light-emission mechanism of Ag-coated ZnO/Al2O3 core/shell nanorod structures.
Journal of nanoscience and nanotechnology (2013-07-19)
Bum-Young Noh, Seong-Ho Baek, Yong-Il Jung, Jae Hyun Kim, Il-Kyu Park
ABSTRACT
Surface plasmon (SP)-enhanced light emission mechanism has been investigated for the Ag-coated ZnO/Al2O3 core/shell nanorods (NRs). Structural characterizations showed that the ZnO NRs were covered by conformal Al2O3 layer and coated by Ag nanoparticles (NPs). The optical studies by photoluminescence (PL) showed abnormal variation of PL intensity with increasing the thickness of Al2O3. For the Ag NPs-coated ZnO NRs without Al2O3 shell layer, the PL emission quenched due to direct transfer of the photo-excited electrical carriers from ZnO NRs to Ag NPs. With thin Al2O3 layers less than 15 nm, the PL intensity increased with increasing the thickness of Al2O3 layers due to weakening of the Förster-type energy transfer while strong SP-mediated PL emission enhancement. For thicker Al2O3 layers than 15 nm, however, the PL intensity decreased with increasing the thickness of Al2O3 layers due to weakening of SP-mediated PL emission enhancement.
Sigma-Aldrich
Gold, foil, thickness 0.05 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 0.1 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, evaporation slug, diam. × L 0.6 cm × 0.6 cm, 99.99% trace metals basis
Sigma-Aldrich
Gold, foil, thickness 0.25 mm, ≥99.9% trace metals basis
Supelco
Aluminum oxide, activated, neutral, Brockmann Activity I
Sigma-Aldrich
Gold, powder, <10 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 0.5 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, rod, diam. 3.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Aluminum oxide, mesoporous, MSU-X (wormhole), average pore size 3.8 nm
Sigma-Aldrich
Gold, powder, <45 μm, 99.99% trace metals basis
Sigma-Aldrich
Gold, foil, thickness 0.5 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, beads, 1-6 mm, 99.999% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 1.0 mm, 99.997% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 0.127 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 0.25 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Zinc oxide, ReagentPlus®, powder, <5 μm particle size, 99.9%
Sigma-Aldrich
Zinc oxide, puriss., meets analytical specification of Ph. Eur., BP, USP, 99-100.5% (calc. for dried substance)
Sigma-Aldrich
Aluminum oxide, standard grade, Brockmann I, activated, basic
Sigma-Aldrich
Aluminum oxide, Brockmann I, standard grade, activated, acidic
Sigma-Aldrich
Aluminum oxide, activated, Brockmann I, standard grade, neutral
Sigma-Aldrich
Aluminum oxide, Type WN-6, Neutral, Activity Grade Super I
Sigma-Aldrich
Aluminum oxide, pore size 58 Å, ~150 mesh
Sigma-Aldrich
Zinc oxide, puriss. p.a., ACS reagent, ≥99.0% (KT)
Sigma-Aldrich
Aluminum oxide, fused, powder, primarily α-phase, -325 mesh
Sigma-Aldrich
Aluminum oxide, powder, 99.99% trace metals basis
Sigma-Aldrich
Aluminum oxide, fused, powder, primarily α-phase, 100-200 mesh
Sigma-Aldrich
Aluminum oxide, Corundum, α-phase, -100 mesh
Sigma-Aldrich
Zinc oxide, dispersion, nanoparticles, <100 nm particle size (TEM), ≤40 nm avg. part. size (APS), 20 wt. % in H2O
Sigma-Aldrich
Aluminum oxide, nanoparticles, <50 nm particle size (DLS), 20 wt. % in isopropanol
Sigma-Aldrich
Gold coated glass cover slip, 99.999% (Au), diam. × thickness 15 mm × 130-170 μm, layer thickness 100 Å