Merck
  • Dehydration of phenylboronic acid to boroxine catalyzed by Au(n) nanoclusters with atom packing core-shell structure.
Dehydration of phenylboronic acid to boroxine catalyzed by Au(n) nanoclusters with atom packing core-shell structure.
Journal of nanoscience and nanotechnology (2013-08-02)
Ping Huang, Zhi Jiang, Guoxiang Chen, Yan Zhu, Yuhan Sun
ABSTRACT
Atomically precise Au(n) nanoclusters (n = number of gold atom in cluster) ideally composed of an exact number of gold atoms have unique core-shell structure and non-metallic electronic properties. The extremely small size of Au25 and Au38 nanoclusters induces a large energy gap in their electronic structures, which gives rise to unprecedented catalytic activity in some chemical reactions. Here we report dehydration of phenylboronic acid to boroxine with small Au25 and Au38 nanocluster catalysts, respectively. Especially, Au25 nanocluster is arranged with Au13 icosahedral core capped by twelve gold atoms as an exterior shell. Au38 nanocluster is a face-fused biicosahedral Au23 core encapsulated by a shell comprised of fifteen gold atoms. Au38 nanoclusters exhibit higher activity than Au25 nanoclusters. This study is an attempt to provide a powerful tool for the catalyst design and to gain a further insight into the correlation of structural properties with catalytic properties.
Product Description
Supelco
Water, for TOC analysis
Sigma-Aldrich
Water, BioPerformance Certified
Supelco
Density Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality
Supelco
Water, ACS reagent, for ultratrace analysis
Supelco
Water, suitable for ion chromatography
Sigma-Aldrich
Gold, foil, thickness 0.05 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 0.1 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, evaporation slug, diam. × L 0.6 cm × 0.6 cm, 99.99% trace metals basis
Sigma-Aldrich
Gold, foil, thickness 0.25 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Gold, powder, <10 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Boron, ≥95% (boron), amorphous powder
Sigma-Aldrich
Gold, wire, diam. 0.5 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, rod, diam. 3.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, powder, <45 μm, 99.99% trace metals basis
Sigma-Aldrich
Gold, foil, thickness 0.5 mm, 99.99% trace metals basis
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Water, Molecular Biology, sterile filtered
Sigma-Aldrich
Water, tested according to Ph. Eur.
Sigma-Aldrich
Gold, beads, 1-6 mm, 99.999% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 1.0 mm, 99.997% trace metals basis
Supelco
Water, suitable for HPCE, suitable for luminescence, suitable for UV/Vis spectroscopy
Sigma-Aldrich
Gold, wire, diam. 0.127 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 0.25 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Boron, crystalline, 1 cm, 99.7% trace metals basis
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Water, ACS reagent
Sigma-Aldrich
Water, Deionized