Merck
  • New conjugated urinary metabolites in intermediate type maple syrup urine disease.
New conjugated urinary metabolites in intermediate type maple syrup urine disease.
Clinica chimica acta; international journal of clinical chemistry (1987-10-30)
L Hagenfeldt, A S Naglo
ABSTRACT
N-Lactylvaline, N-lactylleucine, N-lactylisoleucine and the N-2-hydroxyisovaleryl conjugates of glycine, valine, leucine and isoleucine have been identified in urine from a patient with the intermediate type of maple syrup urine disease.
Sigma-Aldrich
(S)-(+)-2-Hydroxy-3-methylbutyric acid, 99%, optical purity ee: 99% (GLC)
Sigma-Aldrich
2-Hydroxy-3-methylbutyric acid, 99%
Sigma-Aldrich
D-α-Hydroxyisovaleric acid, ≥98.0% (T)