Merck
  • Unexpected adverse effects of Freon 11 and Freon 12 as medication propellants.
Unexpected adverse effects of Freon 11 and Freon 12 as medication propellants.
The Journal of the American Osteopathic Association (1993-06-01)
R J Oenbrink
ABSTRACT
Metered-dose inhalers are frequently used in treating pulmonary diseases associated with bronchoconstriction, chiefly asthma and chronic bronchitis. These aerosolized medications are not without the potential for adverse effects. The author describes two patients who likely had adverse reactions to the Freon propellants used in the inhalers. These reactions are reported in order to alert physicians to their possible occurrence and to suggest a rational treatment approach.
Product Description
Supelco
Trichlorofluoromethane, analytical standard
Sigma-Aldrich
Trichlorofluoromethane, ≥99.5%
Sigma-Aldrich
Trichlorofluoromethane solution, suitable for NMR (reference standard), 10% in acetone-d6 (99.9 atom % D), TMS 6 %, 1,2-dichloro-1,1-difluoroethane 1 %