Merck
  • [Studies on "Fast Green FCF Standard" for the dye standard on the National Institute of Health Sciences].
[Studies on "Fast Green FCF Standard" for the dye standard on the National Institute of Health Sciences].
Eisei Shikenjo hokoku. Bulletin of National Institute of Hygienic Sciences (1996-01-01)
S Ishimitsu, I Mishima, S Tsuji, T Shibata
ABSTRACT
The raw material for Fast Green FCF was tested for preparation of the "Fast Green FCF Standard (C.I. 42053)". Analytical data obtained were as follows: paper chromatography, only one spot is observed; arsenic content, 0.38 microgram/g; chloride content, 0.11%; sulfate content, 3.30%; heavy metals, lead, 8.0 micrograms/g, manganese, 28.1 micrograms/g, and chromium, 1.6 micrograms/g; infrared spectra, 1575 cm-1, 1169 cm-1, and 1033 cm-1; loss on drying, 2.39%; assay, 93.0% by the titanium trichloride titration. Based on the above results, the raw material was authorized as the Dye Standard of National Institute of Health Sciences.
Sigma-Aldrich
Titanium(III) chloride solution, 10-15% TiCl3 basis
Supelco
Fast Green FCF, analytical standard
Sigma-Aldrich
Fast Green FCF, certified by the BSC
Sigma-Aldrich
Fast Green FCF, Dye content ≥85 %