Merck
Response of pigs to bitter-tasting compounds.
Chemical senses (1997-04-01)
S L Nelson, J D Sanregret
ABSTRACT
Two-bottle preference tests were done to determine whether pigs detect bitter-tasting compounds. Four standard bitter compounds and nine bitter-tasting pharmaceutical compounds were tested. Pigs detect and avoid taste compounds that humans perceive as bitter-tasting. A dose-response to varying concentrations of bitter tastants can be measured.
Sigma-Aldrich
Sucrose octaacetate, ≥97%, FG
Sigma-Aldrich
D-(+)-Sucrose octaacetate, 98%