Merck
  • [Antazoline/tetryzoline eyedrops in comparison with levocabastine eyedrops in acute allergic conjunctivitis].
[Antazoline/tetryzoline eyedrops in comparison with levocabastine eyedrops in acute allergic conjunctivitis].
Klinische Monatsblatter fur Augenheilkunde (1997-02-01)
R Wertheimer, G Blessmann
ABSTRACT
Allergic conjunctivitis is one of the most frequent allergic diseases of the anterior eye segment. This multicentre, clinical trial was an investigation to compare the antiallergic efficacy, local tolerance and safety of Antazolin/Tetryzolin eye drops and Levocabastine eye drops. 69 patients were treated over a 2 weeks course of therapy. The subjective and objective ocular symptoms were documented over the treatment period. Both eye drops reduced subjective and objective ocular symptoms effective. The difference between the treatments (p = 0.0395) was the faster onset of action of Antazolin/Tetryzolin 30 minutes after administration of the first drop of trial medication. A fast and effective onset of action is of high clinical relevance. Therefore the benefits of using Antazolin/Tetryzolin eye drops was clearly outweigh.
Sigma-Aldrich
Antazoline hydrochloride