Merck
Identification of the cyclosporin-binding site in P-glycoprotein.
Biochemistry (1999-01-28)
M Demeule, A Laplante, G F Murphy, R M Wenger, R Béliveau
ABSTRACT
The binding site of cyclosporin A to P-glycoprotein was characterized by using a multidrug-resistant Chinese hamster ovary cell line. P-glycoprotein photolabeled with diazirine-cyclosporin A analogue was purified by a two-step process involving continuous elution electrophoresis followed by wheat germ agglutinin-agarose precipitation. The cyclosporin A covalently bound to P-glycoprotein and to subsequent proteolytic fragments was detected by Western blot analysis using a monoclonal antibody against cyclosporin A. Proteolytic digestion of purified P-glycoprotein by V8 generated a major fragment of 15 kDa photolabeled by cyclosporin A, while proteolysis of P-glycoprotein photolabeled by [125I]-iodoaryl azidoprazosin generated a major fragment of 7 kDa. Limited proteolysis of cyclosporin A-photolabeled P-glycoprotein with trypsin indicated that the major binding site for cyclosporin A was in the C-terminal half of the protein. This cyclosporin A binding site was further characterized with chemical agents (N-chlorosuccinimide, cyanogen bromide, and 2-nitro-5-thiocyanobenzoate). These three chemical agents established a proteolytic profile of P-glycoprotein for fragments photolabeled with cyclosporin A and for fragments that contained the C494 and C219 epitopes. The smallest fragments generated by these chemical agents include the transmembrane domains (TMs) 10, 11, and 12 of P-glycoprotein. When the fragments generated by these chemical agents are aligned, the region that binds cyclosporin A is reduced to the 953-1007 residues. These combined results suggest that the major binding site of cyclosporin A occurs between the end of TM 11 and the end of TM 12.
Sigma-Aldrich
N-Chlorosuccinimide, 98%
Sigma-Aldrich
2-Nitro-5-thiocyanatobenzoic acid, powder