Merck
  • Molecular mapping of pulmonary endothelial membrane glycoproteins of the intact rabbit lung.
Molecular mapping of pulmonary endothelial membrane glycoproteins of the intact rabbit lung.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (1990-09-01)
M P Merker, W W Carley, C N Gillis
ABSTRACT
To study the biochemical characteristics of endothelium in vivo, we radioiodinated endothelial membrane proteins of the perfused rabbit lung using a water soluble analog of the Bolton-Hunter reagent, 125I-sulfosuccinimidyl (hydroxyphenyl) propionate (125I-s-SHPP). This technique led to a 10-fold increase in specific activity of radioiodinated lung membrane protein compared with our previously reported method using lactoperoxidase and glucose oxidase-catalyzed radioiodination. Tissue autoradiography confirmed that radioiodination was largely confined to the endothelium. Perfusion pressure, wet-to-dry weight ratios, and the morphological appearance of the lungs were within normal limits, indicating that the procedure does not cause apparent lung injury. Lectin binding to a crude membrane fraction of 125I-s-SHPP labeled lung led to isolation of several putative endothelial membrane proteins. Immunoprecipitation studies with appropriate antibodies enabled the identification of radioiodinated angiotensin-converting enzyme and beta 2-microglobulin associated major histocompatibility complex class I molecules in the membrane fraction. This technique will be useful for studying biochemical responses of the endothelium in vivo to a variety of pharmacological and physiology stimuli.
Sigma-Aldrich
3-(4-Hydroxyphenyl)propionic acid N-hydroxysuccinimide ester, BioReagent, suitable for fluorescence, ≥97% (C)
Sigma-Aldrich
3-(4-Hydroxyphenyl)propionic acid N-hydroxysuccinimide ester, ~90%