- DALDA analogues containing alpha-hydroxymethylamino acids.
DALDA analogues containing alpha-hydroxymethylamino acids.
Acta biochimica Polonica (2002-05-09)
A Olma, N N Chung, P W Schiller, J Zabrocki
PMID11995977
ABSTRACT
To evaluate the role of aromatic amino-acids residues, four analogues of the mu-selective opioid peptide agonist DALDA (H-Tyr-D-Arg-Phe-Lys-NH2) containing the amphiphilic, a,a-disubstituted amino acid (R)- or (S)-alpha-hydroxymethyltyrosine (HmTyr) in position 1 and (R)- or (S)-alpha-hydroxymethylphenylalanine (HmPhe) in position 3 of the peptide sequence were synthesized. Only the [(R)-HmPhe3)]DALDA analogue displayed full agonistic activity in both the guinea pig ileum and the mouse vas deferens assays and turned out to be a delta receptor-selective opioid agonist.