Merck
  • Activation of alpha(2)-adrenoceptors in the lateral hypothalamus reduces pilocarpine-induced salivation in rats.
Activation of alpha(2)-adrenoceptors in the lateral hypothalamus reduces pilocarpine-induced salivation in rats.
Neuroscience letters (2008-12-02)
Ana C Takakura, Thiago S Moreira, Débora S A Colombari, Laurival A De Luca, José V Menani
ABSTRACT
Anti-hypertensive drugs that act on central alpha(2)-adrenoceptors and imidazoline receptors usually cause dry mouth in patients. A central area important for the control of salivary secretion and also for the effects of alpha(2)-adrenoceptor activation is the lateral hypothalamus (LH). Therefore, in the present study we investigated the effects of the injections of moxonidine (an alpha(2)-adrenoceptor and imidazoline agonist) alone or combined with RX 821002 (alpha(2)-adrenoceptor antagonist) into the LH on the salivation induced by intraperitoneal (i.p.) pilocarpine (cholinergic muscarinic agonist). Male Holtzman rats with stainless steel cannula implanted into the LH were used. Saliva was collected using pre-weighted small cotton balls inserted into the animal's mouth under ketamine anesthesia. Salivation induced by i.p. pilorcarpine (4micromol/kg of body weight) was reduced by the injection of moxonidine (10 and 20nmol/0.5microl) into the LH (222+/-46 and 183+/-19mg/7min, vs. vehicle: 480+/-30mg/7min). The inhibitory effect of moxonidine on pilocarpine-induced salivation was abolished by prior injections of RX 821002 (160 and 320nmol/0.5microl) into the LH (357+/-25 and 446+/-38mg/7min). Injections of the alpha(1)-adrenoceptor antagonist prazosin (320nmol/0.5microl) into the LH did not change the effects of moxonidine. The results show that activation of alpha(2)-adrenoceptors in the LH inhibits pilocarpine-induced salivation, suggesting that LH is one of the possible central sites involved in the anti-salivatory effects produced by the treatment with alpha(2)-adrenoceptor agonists.
Sigma-Aldrich
RX 821002 hydrochloride, solid, ≥98% (HPLC)