Merck
  • Enantioselective total synthesis of (+)-lysergic acid, (+)-lysergol, and (+)-isolysergol by palladium-catalyzed domino cyclization of allenes bearing amino and bromoindolyl groups.
Enantioselective total synthesis of (+)-lysergic acid, (+)-lysergol, and (+)-isolysergol by palladium-catalyzed domino cyclization of allenes bearing amino and bromoindolyl groups.
The Journal of organic chemistry (2011-03-03)
Shinsuke Inuki, Akira Iwata, Shinya Oishi, Nobutaka Fujii, Hiroaki Ohno
ABSTRACT
Enantioselective total synthesis of the biologically important indole alkaloids (+)-lysergol, (+)-isolysergol, and (+)-lysergic acid is described. Key features of these total synthesis include (1) a facile synthesis of a chiral 1,3-amino alcohol via the Pd(0)- and In(I)-mediated reductive coupling reaction between L-serine-derived 2-ethynylaziridine and formaldehyde; (2) the Cr(II)/Ni(0)-mediated Nozaki-Hiyama-Kishi (NHK) reaction of an indole-3-acetaldehyde with iodoalkyne; and (3) Pd(0)-catalyzed domino cyclization of an allene bearing amino and bromoindolyl groups. This domino cyclization enabled direct construction of the C/D ring system of the ergot alkaloids skeleton, as well as the creation of the C5 stereogenic center with transfer of the allenic axial chirality to the central chirality.