Merck
  • Inhibition of DNA repair with MGMT pseudosubstrates: phase I study of lomeguatrib in combination with dacarbazine in patients with advanced melanoma and other solid tumours.
Inhibition of DNA repair with MGMT pseudosubstrates: phase I study of lomeguatrib in combination with dacarbazine in patients with advanced melanoma and other solid tumours.
British journal of cancer (2011-08-04)
H A Tawbi, L Villaruz, A Tarhini, S Moschos, M Sulecki, F Viverette, J Shipe-Spotloe, R Radkowski, J M Kirkwood
ABSTRACT
The DNA repair protein O6-methylguanine-DNA methyltransferase (MGMT) reverses the O6-methylguanine (O6-meG) lesion induced by dacarbazine. Depletion of MGMT can be achieved using O6-meG pseudosubstrates. Herein, we report the first phase I experience of the novel O6-meG pseudosubstrate lomeguatrib, combined with dacarbazine. This is a phase I dose-escalation study to determine the maximum tolerated dose and recommended phase II dose (RP2D) of lomeguatrib combined with a single dose of dacarbazine on a 21-day schedule. The vast majority of the 41 patients enrolled had metastatic melanoma (36/41) and most had no previous chemotherapy (30/41). The most frequent non-hematological adverse events (AEs) were nausea (52%), and fatigue (42%). The most frequent AEs of grade 3-4 severity were neutropaenia (42%), leukopaenia (17%), and thrombocytopaenia (12%). Only 1 patient had a partial response and 10 patients had stable disease. The RP2D of lomeguatrib was 40 mg orally twice daily for 10 days combined with 400 mg m(-2) of dacarbazine IV on day 2. Oral administration of lomeguatrib substantially increases the haematological toxicity of dacarbazine consistent with experience with other O6-meG pseudosubstrates.
Sigma-Aldrich
O6-Benzylguanine, ≥98% (TLC), solid
Sigma-Aldrich
Lomeguatrib, ≥98% (HPLC)