Merck
  • Metabolism-directed design of oxetane-containing arylsulfonamide derivatives as γ-secretase inhibitors.
Metabolism-directed design of oxetane-containing arylsulfonamide derivatives as γ-secretase inhibitors.
Journal of medicinal chemistry (2011-10-15)
Antonia F Stepan, Kapil Karki, W Scott McDonald, Peter H Dorff, Jason K Dutra, Kenneth J Dirico, Annie Won, Chakrapani Subramanyam, Ivan V Efremov, Christopher J O'Donnell, Charles E Nolan, Stacey L Becker, Leslie R Pustilnik, Blossom Sneed, Hao Sun, Yasong Lu, Ashley E Robshaw, David Riddell, Theresa J O'Sullivan, Evelyn Sibley, Steven Capetta, Kevin Atchison, Andrew J Hallgren, Emily Miller, Anthony Wood, R Scott Obach
ABSTRACT
A metabolism-based approach toward the optimization of a series of N-arylsulfonamide-based γ-secretase inhibitors is reported. The lead cyclohexyl analogue 6 suffered from extensive oxidation on the cycloalkyl motif by cytochrome P450 3A4, translating into poor human liver microsomal stability. Knowledge of the metabolic pathways of 6 triggered a structure-activity relationship study aimed at lowering lipophilicity through the introduction of polarity. This effort led to several tetrahydropyran and tetrahydrofuran analogues, wherein the 3- and 4-substituted variants exhibited greater microsomal stability relative to their 2-substituted counterparts. Further reduction in lipophilicity led to the potent γ-secretase inhibitor and 3-substituted oxetane 1 with a reduced propensity toward oxidative metabolism, relative to its 2-substituted isomer. The slower rates of metabolism with 3-substituted cyclic ethers most likely originate from reductions in lipophilicity and/or unfavorable CYP active site interactions with the heteroatom. Preliminary animal pharmacology studies with a representative oxetane indicate that the series is generally capable of lowering Aβ in vivo. As such, the study also illustrates the improvement in druglikeness of molecules through the use of the oxetane motif.
Sigma-Aldrich
Trimethylene oxide, 97%