Merck
  • Induction of morphine-6-glucuronide synthesis by heroin self-administration in the rat.
Induction of morphine-6-glucuronide synthesis by heroin self-administration in the rat.
Psychopharmacology (2011-10-22)
Maria Meringolo, Valentina Brusadin, Maria T De Luca, Christian Montanari, Christian L Montanari, Letizia Antonilli, Paolo Nencini, Aldo Badiani
ABSTRACT
Heroin is rapidly metabolized to morphine that in turn is transformed into morphine-3-glucuronide (M3G), an inactive metabolite at mu-opioid receptor (MOR), and morphine-6-glucuronide (M6G), a potent MOR agonist. We have found that rats that had received repeated intraperitoneal injections of heroin exhibit measurable levels of M6G (which is usually undetectable in this species). The goal of the present study was to investigate whether M6G synthesis can be induced by intravenous (i.v.) heroin self-administration (SA). Rats were trained to self-administer either heroin (50 μg/kg per infusion) or saline for 20 consecutive 6-h sessions and then challenged with an intraperitoneal challenge of 10 mg/kg of heroin. Plasma levels of heroin, morphine, 6-mono-acetyl morphine, M3G, and M6G were quantified 2 h after the challenge. In vitro morphine glucuronidation was studied in microsomal preparations obtained from the liver of the same rats. Heroin SA induced the synthesis of M6G, as indicated by detectable plasma levels of M6G (89.7 ± 37.0 ng/ml vs. 7.35 ± 7.35 ng/ml after saline SA). Most important, the in vitro V (max) for M6G synthesis was correlated with plasma levels of M6G (r (2) = 0.78). Microsomal preparations from saline SA rats produced negligible amounts of M6G. Both in vivo and in vitro data indicate that i.v. heroin SA induces the synthesis of M6G. These data are discussed in the light of previous studies conducted in heroin addicts indicating that in humans heroin enhances the synthesis of the active metabolite of heroin and morphine.
Product Description
Supelco
Morphine-6-β-D-glucuronide solution, 100 μg/mL in methanol: water (1:1), ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Morphine-6-β-D-glucuronide solution, 1.0 mg/mL in methanol: water (2:8), ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Morphine-3-β-D-glucuronide solution, 1 mg/mL in methanol with 0.05% NaOH, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Morphine-3-β-D-glucuronide solution, 100 μg/mL in methanol: water ( 50:50(v/v)), ampule of 1 mL, certified reference material, Cerilliant®