Merck
  • Influence of temperature and interactions with ligands on dissociation of dsDNA and ligand-dsDNA complexes of various types of binding. An electrochemical study.
Influence of temperature and interactions with ligands on dissociation of dsDNA and ligand-dsDNA complexes of various types of binding. An electrochemical study.
Physical chemistry chemical physics : PCCP (2012-02-03)
Ewelina Zabost, Anna Maria Nowicka, Zofia Mazerska, Zbigniew Stojek
ABSTRACT
Several medicinally important compounds that bind to dsDNA strands via intercalation (C-1311, C-1305, EtBr), major groove binding (Hoechst 33258) and covalent binding (cis-Pt) were examined. The obtained results suggest that both the transfer of conformation B to C and the denaturation process, for the ligand-dsDNA complexes, except for covalently bound cis-Pt, took place at higher temperatures compared to the unbound helix. Furthermore, much lower currents of electrooxidation of guanine at 100 °C, compared to the currents obtained at this temperature for dsDNA in the absence of ligands, suggest that the binding of ligands affects the way the dsDNA denaturates at increased temperatures and leads to formation of different forms of DNA single strands. The voltammetric results were compared with the data of two spectroscopic techniques: UV-Vis and CD.
Sigma-Aldrich
Symadex, ≥98% (HPLC)