Merck
Bone metastases radiopharmaceuticals: an overview.
Current radiopharmaceuticals (2013-03-09)
Vincenzo Cuccurullo, Giuseppe Lucio Cascini, Oscar Tamburrini, Antonio Rotondo, Luigi Mansi
ABSTRACT
The skeleton is one of the preferential sites for metastases of solid tumors, and metastatic disease is the most common malignancy of the bone. Diagnosis and evaluation of skeletal metastases require more frequently a combined approach of different diagnostic methods. Between the currently available imaging modalities, a major role is devoted to two radionuclide functional techniques namely scintigraphy and positron emission tomography (PET) imaging. Both these techniques require the use of different radiopharmaceuticals. The aim of this paper is to review the most important radiocompounds that can be successfully used to detect and/or characterize bone metastases.
Sigma-Aldrich
Sodium fluoride 0.5 M solution
Supelco
Fluoride ion solution for ISE, 0.1 M F-, for ion-selective electrodes
Sigma-Aldrich
Sodium fluoride, 99.99% trace metals basis
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sigma-Aldrich
Sodium fluoride, BioReagent, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium fluoride, powder, 99.99% trace metals basis
Sigma-Aldrich
Sodium fluoride, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium fluoride, ACS reagent, ≥99%
Sigma-Aldrich
Sodium fluoride, puriss., meets analytical specification of Ph. Eur., BP, USP, 98.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Thymidine, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Thymidine, ≥99%
Sigma-Aldrich
Sodium Fluoride Solution
Sigma-Aldrich
3,4-Dihydroxy-DL-phenylalanine
Sigma-Aldrich
Thymidine, ≥99.0% (HPLC)