Merck
Allergic contact dermatitis to dimethoxane in a spin finish.
Contact dermatitis (1980-10-01)
E Shmunes, R J Kempton
ABSTRACT
A textile worker exposed to dimethoxane in an aqueous system developed allergic contact dermatitis. Patch testing to this compound and its possible products in an aqueous system demonstrated sensitization to acetaldehyde and crotonaldehyde.