Merck
  • Liquid chromatographic-mass spectrometric analysis of N-acetylamino acids in human urine.
Liquid chromatographic-mass spectrometric analysis of N-acetylamino acids in human urine.
Journal of chromatography. B, Biomedical applications (1994-07-01)
K Sugahara, J Zhang, H Kodama
ABSTRACT
Liquid chromatography-atmospheric pressure chemical-ionization mass spectrometry (LC-APCI-MS) was used for the analysis of N-acetylamino acids that could not be determined with an amino acid analyzer. LC-APCI-MS could directly detect the protonated molecular ions of various synthetic N-acetylamino acids, distinguishing N-acetylserine from O-acetylserine and N(alpha)-acetyllysine from N(epsilon)- acetyllysine. Furthermore, N-acetylasparagine, N-acetylaspartic acid, N-acetylglutamine and N-acetylglutamic acid were identified in normal human urine. The assay data for N-acetylaspartic acid agree with a previous report using gas chromatography-mass spectrometry. These results demonstrate the usefulness of the apparatus described above for the analysis of N-acetylamino acids in biological samples.
Sigma-Aldrich
N-Acetyl-L-glutamic acid, ReagentPlus®, 99%
Sigma-Aldrich
Nα-Acetyl-L-asparagine, 98%