Merck
Resveratrol is a selective human cytochrome P450 1A1 inhibitor.
Biochemical and biophysical research communications (1999-08-17)
Y J Chun, M Y Kim, F P Guengerich
ABSTRACT
Resveratrol (trans-3,4',5-trihydroxystilbene) is a phytoalexin compound found in juice and wine produced from dark-skinned grape cultivars and reported to have anti-inflammatory and anticarcinogenic activities. To investigate the mechanism of anticarcinogenic activities of resveratrol, the effects on cytochrome P450 (P450) were determined in human liver microsomes and Escherichia coli membranes coexpressing human P450 1A1 or P450 1A2 with human NADPH-P450 reductase (bicistronic expression system). Resveratrol slightly inhibited ethoxyresorufin O-deethylation (EROD) activity in human liver microsomes with an IC(50) of 1.1 mM. Interestingly, resveratrol exhibited potent inhibition of human P450 1A1 in a dose-dependent manner with IC(50) of 23 microM for EROD and IC(50) of 11 microM for methoxyresorufin O-demethylation (MROD). However, the inhibition of human P450 1A2 by resveratrol was not so strong (IC(50) 1.2 mM for EROD and 580 microM for MROD). Resveratrol showed over 50-fold selectivity for P450 1A1 over P450 1A2. The activities of human NADPH-P450 reductase were not significantly changed by resveratrol. In a human P450 1A1/reductase bicistronic expression system, resveratrol inhibited human P450 1A1 activity in a mixed-type inhibition (competitive-noncompetitive) with a K(i) values of 9 and 89 microM. These results suggest that resveratrol is a selective human P450 1A1 inhibitor, and may be considered for use as a strong cancer chemopreventive agent in humans.
Product Description
Supelco
Resveratrol, analytical standard
Supelco
Resveratrol, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Resveratrol, ≥99% (HPLC)