Merck
  • Effects of aldehyde dehydrogenase-2 genotype on cardiovascular and endocrine responses to alcohol in young Japanese subjects.
Effects of aldehyde dehydrogenase-2 genotype on cardiovascular and endocrine responses to alcohol in young Japanese subjects.
Autonomic neuroscience : basic & clinical (2002-12-21)
Fusae T Nishimura, Tatsushige Fukunaga, Hideaki Kajiura, Katsumi Umeno, Hiromasa Takakura, Taketoshi Ono, Hisao Nishijo
ABSTRACT
The present study was designed to investigate the effects of aldehyde dehydrogenase-2 (ALDH2) genotype on cardiovascular and endocrine responses to alcohol ingestion in young, healthy Japanese subjects. For this purpose, we monitored changes in the electrocardiogram (ECG), blood pressure (BP), finger blood flow (BF) and facial skin temperature (FST) during and after alcohol ingestion (0.4 ml/kg body weight). Spectral analyses of beat-to-beat variations of heart rate (HR), BP and BF were applied. Two major spectral components were examined at low frequency (LF: 0.04-0.15 Hz) and high frequency (HF: 0.15-10.4 Hz) bands for HR and BP variability (BPV). Significant effects for ALDH2 genotype were observed in HR variability (HRV) analysis; HF power of HRV was markedly depressed and the LF/HF ratio was significantly higher with alcohol in ALDH2-deficient (ND) subjects, while ALDH2-normal (NN) subjects did not display such changes. Analysis of BP variability showed increased LF and HF power after alcohol ingestion in the NN subjects, but there were no significant differences between genotypic groups. We also examined BF variability (BFV) in six major spectral components; power of the 0.8-2.2 Hz frequency band was significantly affected by genotype and higher power was observed in the ND subjects. Plasma concentrations of both epinephrine and norepinephrine increased after alcohol ingestion only in the ND subjects. Furthermore, plasma concentrations and urinary excretion of epinephrine, but not norepinephrine, were higher after alcohol ingestion in the ND than in the NN subjects. Blood acetaldehyde levels were about 10-fold higher in the ND than in the NN subjects although blood alcohol levels similarly increased in the ND and NN subjects. Our results also indicated that alcohol ingestion increased secretion of pituitary-adrenal hormones including ACTH, beta-endorphin and cortisol in the ND subjects. The present results along with previous studies suggest that alcohol-induced tachycardia in the ND subjects was probably mediated by acetaldehyde-induced rise in epinephrine secretion from the adrenal medulla and/or changes in the autonomic nervous system. Alcohol-induced relative predominance of cardiac sympathetic activity in the ND subjects might be ascribed partly to increased norepinephrine secretion from sympathetic nerve terminals. Effects of acetaldehyde on these cardiovascular and endocrine systems were discussed in terms of their effects on the central nervous system.
Sigma-Aldrich
Acetaldehyde, ACS reagent, ≥99.5%
Supelco
Acetaldehyde, PESTANAL®, analytical standard
Sigma-Aldrich
Acetaldehyde, natural, FG
Sigma-Aldrich
Acetaldehyde, puriss. p.a., anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Acetaldehyde, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Acetaldehyde solution, natural, 50 wt. % ethanol, FG
Sigma-Aldrich
Acetaldehyde solution, 50 wt. % in ethanol
Sigma-Aldrich
Acetaldehyde solution, 40 wt. % in H2O
Sigma-Aldrich
Acetaldehyde solution, 40 wt. % in isopropanol
Sigma-Aldrich
Acetaldehyde solution, 5 M in THF