Merck
  • Metabolite profile of cerebrospinal fluid in patients with spina bifida: a proton magnetic resonance spectroscopy study.
Metabolite profile of cerebrospinal fluid in patients with spina bifida: a proton magnetic resonance spectroscopy study.
Spine (2005-02-01)
Kamalesh Pal, Uma Sharma, D K Gupta, Akshay Pratap, N R Jagannathan
ABSTRACT
The present study was carried out to assess the metabolic differences between cerebrospinal fluid samples of patients with spina bifida and age-matched control individuals. To study the metabolite profile of cerebrospinal fluid of patients with spina bifida using proton magnetic resonance spectroscopy, compare the levels of metabolites with controls, establish correlation of underlying neuronal dysfunction with metabolic changes in patients with spina bifida, and evaluate the potential use of this technique as an additional tool for diagnostic assessment. Combination of embryopathy, stretching, ischemia, compression, and trauma is responsible for cord dysfunction in spina bifida. Changes in neuronal metabolism leads to changes in the local milieu of cerebrospinal fluid in the cord. Change in metabolite profile of cerebrospinal fluid in spina bifida in terms of increase in products of anaerobic metabolism, nerve membrane integrity, and nerve ischemia has not yet been studied. Cerebrospinal fluid obtained from patients and control individuals were characterized using various one- and two-dimensional proton magnetic resonance spectroscopy techniques. Concentration of various metabolites was calculated using the area under the nuclear magnetic resonance peak. Statistically significantly higher levels of lactate, choline, glycerophosphocholine, acetate, and alanine in the cerebrospinal fluid of patients with spina bifida was observed compared with control individuals. Significantly higher levels of metabolites were observed in patients with spina bifida, representing a state of nerve ischemia, anaerobic metabolism, and disruption of neuronal membrane.
Sigma-Aldrich
4-Hydroxyphenylacetic acid, 98%