  • Search Results
  • In vivo characterization of 123/125I-2-iodo-L-phenylalanine in an R1M rhabdomyosarcoma athymic mouse model as a potential tumor tracer for SPECT.
In vivo characterization of 123/125I-2-iodo-L-phenylalanine in an R1M rhabdomyosarcoma athymic mouse model as a potential tumor tracer for SPECT.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine (2005-03-08)
Veerle Kersemans, Bart Cornelissen, Ken Kersemans, Matthias Bauwens, Erik Achten, Rudi A Dierckx, John Mertens, Guido Slegers
ABSTRACT
The application of 123I-3-iodo-alpha-methyltyrosine is limited to diagnosis of brain tumors due to its marked long-term uptake in kidneys. In vitro evaluation of 125I-2-iodo-L-phenylalanine showed high uptake in R1M cells by L-type amino acid transport system 1 (LAT1). This study evaluates 123I-2-iodo-L-phenylalanine as a new specific tumor tracer for SPECT. 123/125I-2-Iodo-L-phenylalanine is prepared as a one-pot kit using the Cu1+-assisted isotopic exchange method. The characteristics of 125I-2-iodo-L-phenylalanine were examined in vivo in R1M tumor-bearing athymic mice and in acute inflammation-bearing NMRI mice. The uptake of 123/125I-2-iodo-L-phenylalanine in tumor and other organs of interest was measured by dynamic planar imaging (DPI) and gamma-counting after dissection. Displacement of 123I-2-iodo-L-phenylalanine radioactivity by L-phenylalanine, L-methionine, and L-cysteine was measured. 123I-Iodo-human serum albumin planar imaging was performed to correct for blood-pool activity and MRI was performed to delineate the tumor in DPI. 18F-FDG uptake was measured with an animal PET scanner. 125I-2-Iodo-L-phenylalanine and 18F-FDG uptake in inflamed muscle were compared. 123/125I-2-Iodo-L-phenylalanine showed a high and fast tumor uptake and followed a reversible first-order pattern allowing calculation of the half-life and the time to reach equilibrium (t(R)). Net tumor-to-background ratios up to 6.7 at 60 min were obtained. This radioactivity was significantly displaced by L-phenylalanine, L-methionine, and L-cysteine, pointing to reversible LAT transport. When plotting t(R) of the tumor uptake as a function of tumor volume, a rectangular hyperbolic curve was obtained. The almost constant t(R) values at higher tumor volumes (>4 mL) could be linked to increased necrotic tissue. Fast blood clearance of the tracer through the kidneys to the bladder and low tracer activity in the abdomen and brain were observed. The inflamed muscle showed only a slight increase of 125I-2-iodo-L-phenylalanine uptake (inflammation-to-background ratio, RIB = 1.30 +/- 0.02), in contrast to the high 18F-FDG uptake (RIB = 11.1 +/- 1.7). The in vivo stability of 123/125I-2-iodo-L-phenylalanine was good: Only 7% of free radioiodide and no other labeled metabolites were observed after 90 min. 123/125I-2-Iodo-L-phenylalanine is quickly taken up by the overexpressed LAT1 system in R1M tumors with high tumor specificity. The availability of a kit and the specificity of the tracer make 123I-2-iodo-L-phenylalanine a promising tool for oncologic SPECT.
Sigma-Aldrich
L-Cysteine, ≥97%, FG
Sigma-Aldrich
L-Phenylalanine, 99%, FCC
Sigma-Aldrich
L-Phenylalanine, 99%
Sigma-Aldrich
L-Cysteine, 97%
Sigma-Aldrich
L-Phenylalanine, BioUltra, ≥99.0% (NT)
SAFC
L-Methionine
SAFC
L-Cysteine
Sigma-Aldrich
L-Cysteine, BioUltra, ≥98.5% (RT)
Sigma-Aldrich
L-Cysteine, from non-animal source, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
L-Phenylalanine, reagent grade, ≥98%
Sigma-Aldrich
L-Phenylalanine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Methionine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
L-Methionine, reagent grade, ≥98% (HPLC)
Supelco
L-Methionine, certified reference material, TraceCERT®
Supelco
L-Cysteine, certified reference material, TraceCERT®
Supelco
L-Phenylalanine, analytical standard, for Nitrogen Determination According to Kjeldahl Method
Supelco
L-Phenylalanine, certified reference material, TraceCERT®
Methionine, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Methionine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Phenylalanine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Cysteine, produced by Wacker Chemie AG, Burghausen, Germany, ≥98.0%
Sigma-Aldrich
L-Methionine, BioUltra, ≥99.5% (NT)