  • Search Results
  • Specific transport of S-nitrosocysteine in human red blood cells: Implications for formation of S-nitrosothiols and transport of NO bioactivity within the vasculature.
Specific transport of S-nitrosocysteine in human red blood cells: Implications for formation of S-nitrosothiols and transport of NO bioactivity within the vasculature.
FEBS letters (2005-07-19)
Jörg Sandmann, Kathrin S Schwedhelm, Dimitrios Tsikas
ABSTRACT
The transport of various S-nitrosothiols, NO and NO donors in human red blood cells (RBC) and the formation of erythrocytic S-nitrosoglutathione were investigated. Of the NO species tested only S-nitrosocysteine was found to form S-nitrosoglutathione in the RBC cytosol. L-Serine, L-cysteine and L-lysine inhibited formation of S-nitrosoglutathione. Incubation of RBC pre-incubated with S-[15N]nitroso-L-cysteine with native plasma or platelet-rich plasma led to formation of S-[15N]nitrosoalbumin and inhibited platelet aggregation, respectively. The specific transporter system of S-nitroso-L-cysteine in the RBC membrane may have implications for formation of S-nitrosoalbumin and S-nitrosohemoglobin and for transport of NO bioactivity within the vasculature.
Sigma-Aldrich
L-Cysteine, ≥97%, FG
Sigma-Aldrich
L-Cysteine, 97%
SAFC
L-Cysteine
Sigma-Aldrich
L-Cysteine, BioUltra, ≥98.5% (RT)
Sigma-Aldrich
L-Cysteine, from non-animal source, BioReagent, suitable for cell culture, ≥98%
Supelco
L-Cysteine, certified reference material, TraceCERT®
Sigma-Aldrich
L-Cysteine, produced by Wacker Chemie AG, Burghausen, Germany, ≥98.0%