Merck
  • O-demethylation of epipodophyllotoxins is catalyzed by human cytochrome P450 3A4.
O-demethylation of epipodophyllotoxins is catalyzed by human cytochrome P450 3A4.
Molecular pharmacology (1994-02-01)
M V Relling, J Nemec, E G Schuetz, J D Schuetz, F J Gonzalez, K R Korzekwa
ABSTRACT
We previously demonstrated that O-demethylation of the pendant dimethoxyphenol ring of epipodophyllotoxins to produce their respective catechol metabolites is catalyzed by cytochrome(s) P450 in human liver microsomes. Our objective was to identify the specific human cytochrome(s) P450 responsible for catechol formation. Using a panel of prototypical substrates and inhibitors for specific cytochromes P450, we identified substrates for CYP3A4 (midazolam, erythromycin, cyclosporin, and dexamethasone) as inhibitors of catechol formation from both etoposide and teniposide. Dexamethasone inhibition was competitive, with Ki values of 60 and 45 microM for etoposide and teniposide, respectively. In 58 human livers, the correlation coefficients for teniposide catechol formation versus 1'- and 4-hydroxymidazolam formation were 80% and 85%, respectively; for etoposide catechol formation versus 1'- and 4-hydroxymidazolam formation r2 was 83% and 79%, respectively. Teniposide and etoposide catechol formation rates were also significantly correlated with immunodetectable CYP3A (r2 = 49% and 51%, respectively) and not with immunodetectable CYP1A2, 2E1, or 2C8. Finally, cDNAs for human CYP3A4, 3A5, 2A6, 2B6, 2C8, and 2C9 were functionally expressed in HepG2 cells, using a vaccinia viral vector. Teniposide and etoposide catechol formation was catalyzed primarily by 3A4 (15.4 and 40.9 pmol/pmol/hr, respectively) and to a lesser degree by 3A5 (1.94 and 11.3 pmol/pmol/hr, respectively), whereas there was no detectable O-demethylation of epipodophyllotoxins by 2A6, 2B6, 2C8, 2C9, or the control virus alone. Moreover, the relative activities of midazolam hydroxylation, compared with O-demethylation of epipodophyllotoxins, were similar for heterologously expressed 3A4 and for human liver microsomes. We conclude that catechol formation from teniposide and etoposide is primarily mediated by human CYP3A4, making these reactions susceptible to inhibition by prototypical 3A substrates and inhibitors.
Sigma-Aldrich
Pyrocatechol, purified by sublimation, ≥99.5%
Sigma-Aldrich
Pyrocatechol, ≥99%
Sigma-Aldrich
1,2-Dihydroxybenzene, ReagentPlus®, ≥99%