Merck
  • Laser induced resonance energy transfer--a novel approach towards achieving high sensitivity in capillary electrophoresis. I. Clinical diagnostic application.
Laser induced resonance energy transfer--a novel approach towards achieving high sensitivity in capillary electrophoresis. I. Clinical diagnostic application.
Journal of chromatography. A (1996-09-13)
J R Petersen, M G Bissell, A A Mohammad
ABSTRACT
Most of the procedures currently performed by capillary zone electrophoresis (CZE) with laser induced fluorescence detection requires prior derivatization. This increases cost, the turn-around-time and chances of extraneous contaminations. CZE with laser induced resonance energy transfer is demonstrated as a viable alternative for detecting non-fluorescent compounds with no prior derivatization. The feasibility of this approach is demonstrated by separating and directly detecting salicylic acid (2,4-dihydroxybenzoic acid), gentisic acid (o-methoxybenzoic acid), salicyluric acid (o-hydroxyhippuric acid) and 4-aminosalicylic acid in urine. The detection of salicylate in serum is also shown. The method is highly sensitive with detection limits in the 1.10(-7) M range. Importantly it requires no prior preconcentration or sample preparation and can be used with complex sample matrices such as serum and urine.
Product Description
Supelco
3,4-Dihydroxybenzoic acid, analytical standard
Protocatechuic acid, primary reference standard
Sigma-Aldrich
3,4-Dihydroxybenzoic acid, ≥97.0% (T)