Merck
  • Effects and action mechanisms of Korean pear (Pyrus pyrifolia cv. Shingo) on alcohol detoxification.
Effects and action mechanisms of Korean pear (Pyrus pyrifolia cv. Shingo) on alcohol detoxification.
Phytotherapy research : PTR (2012-03-28)
Ho-Sun Lee, Toyoshi Isse, Toshihiro Kawamoto, Hyun-Su Woo, An Keun Kim, Jong Y Park, Mihi Yang
ABSTRACT
Korean pear (Pyrus pyrifolia cv. Shingo) has been used as a traditional medicine for alleviating alcohol hangover. However, scientific evidence for its effectiveness or mechanism is not clearly established. To investigate its mechanism of alcohol detoxification, both in vitro and in vivo studies were performed with an aldehyde dehydrogenase 2 (ALDH2) alternated animal model. The pear extract (10 mL/kg bw) was administered to Aldh2 normal (C57BL/6) and deficient (Aldh2 -/-) male mice. After 30 min, ethanol (1 g or 2 g/kg bw) was administered to the mice via gavage. Levels of alcohol and acetaldehyde in blood were quantified by GC/MS. First, it was observed that the pears stimulated both alcohol dehydrogenase (ADH) and ALDH activities by 2∼3-  and 1.3-fold in in vitro studies, respectively. Second, mouse PK data (AUC(∞) and C(max) ) showed that the pear extract decreased the alcohol level in blood regardless of ALDH2 genotype. Third, the pear increased the acetaldehyde level in blood in Aldh2 deficient mice but not in Aldh2 normal mice. Therefore, the consistent in vitro and in vivo data suggest that Korean pears stimulate the two key alcohol-metabolizing enzymes. These stimulations could be the main mechanism of the Korean pear for alcohol detoxification. Finally, the results suggest that polymorphisms of human ALDH2 could bring out individual variations in the effects of Korean pear on alcohol detoxification.
Sigma-Aldrich
Aldehyde Dehydrogenase, potassium-activated from baker′s yeast (S. cerevisiae), lyophilized powder, ≥2.0 units/mg protein
Pricing and availability is not currently available.
Sigma-Aldrich
Alcohol Dehydrogenase from Saccharomyces cerevisiae, ≥300 units/mg protein, lyophilized powder (contains buffer salts), Mw 141-151 kDa
Pricing and availability is not currently available.