Merck
  • Identification of DELE, a novel DAP3-binding protein which is crucial for death receptor-mediated apoptosis induction.
Identification of DELE, a novel DAP3-binding protein which is crucial for death receptor-mediated apoptosis induction.
Apoptosis : an international journal on programmed cell death (2010-06-22)
Tanenobu Harada, Atsushi Iwai, Tadaaki Miyazaki
ABSTRACT
Death associated protein 3 (DAP3) is known to be a highly conserved protein, and is responsible for regulating apoptosis induced by various stimuli. To understand the molecular mechanism of how DAP3 induces apoptosis, we performed yeast two-hybrid screening, and identified a novel DAP3-binding protein termed death ligand signal enhancer (DELE). In this report, we show that DELE actually binds to DAP3 in mammalian cells. We found that the cells stably expressing DELE are susceptible to apoptosis induction by the stimulation of TNF-α and TRAIL. In addition, knockdown of DELE expression rescued the HeLa cells from apoptosis induction by these stimuli. Moreover, activation of caspase-3, caspase-8 and caspase-9 induced by stimulation of TNF-α, anti-Fas or TRAIL was significantly inhibited by the knockdown of DELE expression. These results demonstrated the biological significance of DELE for apoptosis signal mediated by death receptors.
Sigma-Aldrich
Anti-DELE antibody produced in rabbit, affinity isolated antibody