Merck
  • An efficient synthesis of N-tert-butoxycarbonyl-O-cyclohexyl-L-tyrosine.
An efficient synthesis of N-tert-butoxycarbonyl-O-cyclohexyl-L-tyrosine.
Chemical & pharmaceutical bulletin (2001-02-24)
Y Nishiyama, S Ishizuka, S Shikama, K Kurita
ABSTRACT
A facile and efficient synthesis of N-tert-butoxycarbonyl-O-cyclohexyl-L-tyrosine [Boc-Tyr(Chx)-OH] is described. Boc-Tyr-OH was treated with NaH in dimethylformamide and then with 3-bromocyclohexene to give N-Boc-O-(cyclohex-2-enyl)-L-tyrosine [Boc-Tyr(Che)-OH] in 70% yield. Hydrogenation of Boc-Tyr(Che)-OH over PtO2 afforded Boc-Tyr(Chx)-OH in almost quantitative yield. The highest yield was achieved when a side product in the synthesis of Boc-Tyr-OH, Boc-Tyr(Boc)-OH, was not removed, because it was also converted to Boc-Tyr(Che)-OH without any additional manipulations. The new synthetic method described here is convenient for practical use, and would facilitate the widespread use of the Chx group for the hydroxy-protection of Tyr.
Sigma-Aldrich
3-Bromocyclohexene, technical grade, 90%
Pricing and availability is not currently available.