Merck
  • A new strategy for determination of hydroxylamine and phenol in water and waste water samples using modified nanosensor.
A new strategy for determination of hydroxylamine and phenol in water and waste water samples using modified nanosensor.
Environmental science and pollution research international (2013-04-24)
Roya Sadeghi, Hassan Karimi-Maleh, Mohammad A Khalilzadeh, Hadi Beitollahi, Zahra Ranjbarha, Mohammad Bagher Pasha Zanousi
ABSTRACT
A carbon paste electrode modified with p-chloranil and carbon nanotubes was used for the sensitive and selective voltammetric determination of hydroxylamine (HX) and phenol (PL). The oxidation of HX at the modified electrode was investigated by cyclic voltammetry (CV), chronoamperommetry, and electrochemical impedance spectroscopy. The values of the catalytic rate constant (k), and diffusion coefficient (D) for HX were calculated. Square wave voltammetric peaks current of HX and PL increased linearly with their concentrations at the ranges of 0.1-172.0 and 5.0-512.0 μmol L(-1), respectively. The detection limits for HX and PL were 0.08 and 2.0 μmol L(-1), respectively. The separation of the anodic peak potentials of HX and PL reached to 0.65 V, using square wave voltammetry. The proposed sensor was successfully applied for the determination of HX and PL in water and wastewater samples.
Product Description
Supelco
Phenol solution, certified reference material, 500 μg/mL in methanol
Pricing and availability is not currently available.
Supelco
Phenol solution, 5000 μg/mL in methanol, certified reference material
Pricing and availability is not currently available.
Sigma-Aldrich
Hydroxylamine solution, 50 wt. % in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Phenol, Molecular Biology
Pricing and availability is not currently available.
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Phenol solution, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, Molecular Biology
Pricing and availability is not currently available.
Sigma-Aldrich
Phenol solution, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2, BioReagent, Molecular Biology
Pricing and availability is not currently available.
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Phenol, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Phenol, ACS reagent, ≥99.0%
Pricing and availability is not currently available.
Supelco
Phenol, PESTANAL®, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Pricing and availability is not currently available.
Sigma-Aldrich
Hydroxylamine solution, 50 wt. % in H2O, 99.999%
Pricing and availability is not currently available.
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Phenol, puriss., ≥99.5% (GC), meets analytical specification of Ph. Eur., BP, USP, crystalline (detached)
Pricing and availability is not currently available.
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Phenol, BioUltra, Molecular Biology, TE-saturated, ~73% (T)
Pricing and availability is not currently available.
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Pricing and availability is not currently available.
Supelco
Phenol solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Phenol, BioUltra, Molecular Biology, ≥99.5% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Pricing and availability is not currently available.
Supelco
Phenol, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.