Merck
  • Synthesis and X-ray crystal structure of isomeric pyridine-based leuco-TAM dyes, 2,2-(2-(pyridinyl)propane-1,3-diylidene)bis(5-chloro-1,3,3-trimethyl indoline) derivatives and unusual stability of 4-pyridinyl compound.
Synthesis and X-ray crystal structure of isomeric pyridine-based leuco-TAM dyes, 2,2-(2-(pyridinyl)propane-1,3-diylidene)bis(5-chloro-1,3,3-trimethyl indoline) derivatives and unusual stability of 4-pyridinyl compound.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2013-06-06)
So-Young Ma, Sam-Rok Keum
ABSTRACT
The solid-state structures and unequivocal stereochemistries of isomeric pyridine-based leuco-triarylmethane (LTAM) dyes 2,2'-(2-(pyridin-4 or 3-yl)propane-1,3-diylidene)bis(5-chloro-1,3,3-trimethylindoline) derivatives were established by X-ray crystallography. Surprisingly, the EE isomer was formed for the 4-pyridinyl compound, whereas the 3-pyridinyl compound formed ZE isomers. In addition, the latter have a so-called three-bladed propeller conformation, whereas the former possess a Y-shaped conformation. These pyridine-based LTAM compounds stack to form a dimer, adopting either an orthorhombic, with Pcmn space group, or monoclinic crystal system with P21/n space group in the crystal unit cell.
Product Description
Supelco
Pyridine, analytical standard
Sigma-Aldrich
Pyridine hydrobromide, 98%
Sigma-Aldrich
Pyridine, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Pyridine, ≥99%
Sigma-Aldrich
Pyridine, biotech. grade, ≥99.9%
Sigma-Aldrich
Pyridine, ACS reagent, ≥99.0%
Supelco
Pyridine solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Pyridine hydrochloride, 98%
Sigma-Aldrich
Pyridine hydrochloride, purum, ≥98.0% (AT)