Merck
Nocturnal home hemodialysis for a patient with type 1 hyperoxaluria.
American journal of kidney diseases : the official journal of the National Kidney Foundation (2013-07-09)
Troy J Plumb, Melissa L Swee, Jennifer A Fillaus
ABSTRACT
Type 1 primary hyperoxaluria is a genetic disorder caused by deficiency of the liver-specific peroxisomal enzyme alanine-glyoxylate aminotransferase. This enzyme deficiency leads to excess oxalate production and deposition of calcium oxalate salts, resulting in kidney failure and systemic oxalosis. Aside from combined liver/kidney transplantation, no curative treatment exists. Various strategies for optimizing dialysis treatment have been evaluated, but neither conventional hemodialysis nor peritoneal dialysis can keep pace with oxalate production in this patient population. In this report, we describe a patient with end-stage renal disease from type 1 primary hyperoxaluria managed with nocturnal home hemodialysis. Performing hemodialysis 8-10 hours each night with blood flow of 350 mL/min and total dialysate volume of 60 L, she has maintained pre- and postdialysis serum oxalate levels at or below the level of supersaturation. We also review published literature regarding oxalate removal in various modalities of dialysis in patients with type 1 primary hyperoxaluria. In our patient, nocturnal hemodialysis has controlled serum oxalate levels better than conventional hemodialysis therapies. Home nocturnal hemodialysis should be considered an option for management of patients with end-stage renal disease from type 1 hyperoxaluria who are awaiting transplantation.
Sigma-Aldrich
Pyridoxine, ≥98%
Pyridoxine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Supelco
Pyridoxine Hydrochloride (B6), analytical standard
Sigma-Aldrich
Pyridoxine hydrochloride, ≥98% (HPLC)
Sigma-Aldrich
Pyridoxine hydrochloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture
Supelco
Pyridoxine hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Pyridoxine hydrochloride, meets USP testing specifications